missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl 2-(1,3-Dithiole)phosphonate 97.0+%, TCI America™
Supplier: TCI America D39811G
Specifications
| Dimethyl 2-(1,3-Dithiole)phosphonate | |
| C5H9O3PS2 | |
| 2-(1,3-Dithiole)phosphonic Acid Dimethyl Ester | |
| COP(=O)(C1SC=CS1)OC | |
| 212.218 | |
| 212.22 | |
| Liquid |
| 133113-76-5 | |
| 1 g | |
| ZQRMTDYBPMYORJ-UHFFFAOYSA-N | |
| 2-dimethoxyphosphoryl-1,3-dithiole | |
| 10921801 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 133113-76-5 | |
| 212.218 | |
| 2-(1,3-Dithiole)phosphonic Acid Dimethyl Ester | |
| 2-dimethoxyphosphoryl-1,3-dithiole |
| C5H9O3PS2 | |
| ZQRMTDYBPMYORJ-UHFFFAOYSA-N | |
| 10921801 | |
| COP(=O)(C1SC=CS1)OC |
Safety and Handling
TSCA : No
Recommended Storage : Freezer