missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 2-(1,3-Dithiole)phosphonate 97.0+%, TCI America™

Catalog No. p-7128032
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39811G 1 g
D39815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39811G Supplier TCI America Supplier No. D39811G
Only null left

Chemical Identifiers

CAS 133113-76-5
Molecular Formula C5H9O3PS2
Molecular Weight (g/mol) 212.218
InChI Key ZQRMTDYBPMYORJ-UHFFFAOYSA-N
Synonym 2-(1,3-Dithiole)phosphonic Acid Dimethyl Ester
PubChem CID 10921801
IUPAC Name 2-dimethoxyphosphoryl-1,3-dithiole
SMILES COP(=O)(C1SC=CS1)OC

Specifications

Quantity 1 g
Formula Weight 212.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl 2-(1,3-Dithiole)phosphonate
TSCA No
Recommended Storage Freezer