accessibility menu, dialog, popup

UserName

CAS RN 133113-76-5

IUPAC Name: dimethyl (2H-1,3-dithiol-2-yl)phosphonate
Synonyms: dimethyl 2H-1,3-dithiol-2-ylphosphonate
Molecular Weight (g/mol): 212.22
Molecular Formula: C5H9O3PS2
InChi Key: ZQRMTDYBPMYORJ-UHFFFAOYSA-N
SMILES: COP(=O)(OC)C1SC=CS1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results