missing translation for 'onlineSavingsMsg'
Learn More

GVS™ Dilution Bottle Butterfield's Buffer (72Pack)

Catalog No. 10486194
Change view
Click to view available options
Quantity:
72 x 90 mL
72 x 99 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10486194 72 x 99 mL
10486195 72 x 90 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10486194 Supplier GVS™ Supplier No. 10498503
Only null left

Butterfield’s Buffer in 90mL or 99mL prefilled sterile dilution bottles for easy 1:10 dilutions. Used extensively in the food, dairy, and pharmaceutical industries as a general diluent. Contains monobasic potassium phosphate.

Description

  • contains monobasic potassium phosphate and is used extensively in the food, dairy, and pharmaceutical industries. Offered in 90 ml and 99 ml volumes for easy 1:10 and 1:100 dilutions. Recommended as a general diluent in laboratory procedures by the Federal Drug Administrations and in the Bacteriological Analytical Manual. This product is prepared according to Standard Methods for the Examination of Water and Wastewater for use in water testing.
  • Prefilled sterile dilution bottles are designed for sample dilution of water, dairy products, foods, and pharmaceuticals prior to microbiological testing. Final pH for all solutions is 7.2 pH ±0.2 pH at 25°C.
  • Easy open, flip-top, plastic container with a tamper-evident seal.

Features and Benefits

  • Pre-filled and Sterile.
  • Tamper evident sterility strip is integrated as part of the cap. It is much easier to open and use. Never hassle with shrink bands again
  • Room temperature Storage
  • Long shelf life
  • Chemically inert
  • Resistant to sample

Applications

  • For sample dilution of water, dairy products, foods and pharmaceuticals prior to microbiological testing.
  • Used extensively in the food, dairy and pharmaceutical industries.

See 'GVS Life Sciences Microbiology Testing mpg YouTube' Video for further details on use.

Specifications

Chemical Name or Material Butterfield's Buffer
pH 7.2
Quantity 72 x 99 mL