missing translation for 'onlineSavingsMsg'
Learn More

Diluted Hydrochloric Acid, 10% (w/v), NF, 9.5-150%, Spectrum™ Chemical

Catalog No. 18602224
Change view
Click to view available options
Quantity:
500 mL
2.5 L
6 x 500 mL
20 L
4 x 2.5 L
200 L
Packaging:
Amber Glass Bottle
Poly Drum
Poly-Coated Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602224 500 mL Amber Glass Bottle
18602222 2.5 L Poly-Coated Glass Bottle
18602225 6 x 500 mL Amber Glass Bottle
18602223 4 x 2.5 L Poly-Coated Glass Bottle
18602226 20 L Poly Drum
18602227 200 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18602224 Supplier Spectrum Chemical Mfg Cor Supplier No. HY105500MLGL

HY105, 7647-01-0, HCl

Spectrum™ Chemical Diluted Hydrochloric Acid, 10 Percent (w/v), NF is a pH acidifier excipient in pharmaceutical preparations. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

CAS Max % 10%, 90%
Packaging Amber Glass Bottle
Quantity 500 mL
UN Number UN1789
Percent Purity 9.5 to 10.5%
Grade NF
Chemical Name or Material Hydrochloric Acid