missing translation for 'onlineSavingsMsg'
Learn More

Diisononyl Phthalate, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC S1559

Catalog No. S-1559


Only null left
Explore available promotions

Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Diisononyl phthalate28553-12-01000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Diisononyl phthalate
28553-12-0
Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
C26H42O4
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
HBGGXOJOCNVPFY-UHFFFAOYSA-N
418.62
2mL Amber Prescored Glass Ampule
0.1
0.1
1000 μg/mL
Certified Reference Material
Acetone, 67-64-1, 99.9%
1 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only