missing translation for 'onlineSavingsMsg'
Learn More

Diisodecyl Phthalate (mixture of branched chain isomers), TCI America™

Catalog No. P0299500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0299500ML 500 mL
P029925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0299500ML Supplier TCI America Supplier No. P0299500ML
Only null left

Chemical Identifiers

CAS 26761-40-0
Molecular Formula C28H46O4
Molecular Weight (g/mol) 446.672
MDL Number MFCD00048389
InChI Key ZVFDTKUVRCTHQE-UHFFFAOYSA-N
Synonym Phthalic Acid Diisodecyl Ester, DIDP
PubChem CID 33599
ChEBI CHEBI:34709
IUPAC Name bis(8-methylnonyl) benzene-1,2-dicarboxylate
SMILES CC(C)CCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCC(C)C

Specifications

Color Yellow
Quantity 500 mL
Formula Weight 446.67
Physical Form Liquid
Chemical Name or Material Diisodecyl Phthalate (mixture of branched chain isomers)
EINECSNumber (3)-1307
RTECSNumber TI1270000
TSCA Yes