accessibility menu, dialog, popup

UserName

CAS RN 26761-40-0

H3CCH3OOOOCH3CH3

CAS RN 26761-40-0

IUPAC Name: 1,2-bis(8-methylnonyl) benzene-1,2-dicarboxylate
Synonyms: diisodecyl phthalate Didp palatinol Z didp (plasticizer) sicol 184 bis(isodecyl)phthalate plasticized ddp vestinol DZ di-''isodecyl'' phthalate
Molecular Weight (g/mol): 446.67
Molecular Formula: C28H46O4
InChi Key: ZVFDTKUVRCTHQE-UHFFFAOYSA-N
SMILES: CC(C)CCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCC(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results