missing translation for 'onlineSavingsMsg'
Learn More

Diisobutyl Phthalate 98.0+%, TCI America™

Supplier:  TCI America P029825G

 View more versions of this product

Catalog No. P029825G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Diisobutyl Phthalate
Yellow
C16H22O4
25 g
diisobutyl phthalate, palatinol ic, dibp, isobutyl phthalate, hexaplas m/1b, kodaflex dibp, di-iso-butyl phthalate, phthalic acid, diisobutyl ester, phthalic acid diisobutyl ester, di i-butyl phthalate
CC(C)COC(=O)C1=CC=CC=C1C(=O)OCC(C)C
278.35
CHEBI:79053
≥98.0% (GC)
84-69-5
320°C
MFCD00026480
3082
MGWAVDBGNNKXQV-UHFFFAOYSA-N
1,2-bis(2-methylpropyl) benzene-1,2-dicarboxylate
6782
278.35
Liquid

Chemical Identifiers

84-69-5
278.35
MGWAVDBGNNKXQV-UHFFFAOYSA-N
6782
1,2-bis(2-methylpropyl) benzene-1,2-dicarboxylate
C16H22O4
MFCD00026480
diisobutyl phthalate, palatinol ic, dibp, isobutyl phthalate, hexaplas m/1b, kodaflex dibp, di-iso-butyl phthalate, phthalic acid, diisobutyl ester, phthalic acid diisobutyl ester, di i-butyl phthalate
CHEBI:79053
CC(C)COC(=O)C1=CC=CC=C1C(=O)OCC(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1303

RTECSNumber : TI1225000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions