missing translation for 'onlineSavingsMsg'
Learn More

Diisobutyl Phthalate 98.0+%, TCI America™

Catalog No. p-7134542
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P029825G 25 g
P0298500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P029825G Supplier TCI America Supplier No. P029825G
Only null left

Chemical Identifiers

CAS 84-69-5
Molecular Formula C16H22O4
Molecular Weight (g/mol) 278.35
MDL Number MFCD00026480
InChI Key MGWAVDBGNNKXQV-UHFFFAOYSA-N
Synonym diisobutyl phthalate, palatinol ic, dibp, isobutyl phthalate, hexaplas m/1b, kodaflex dibp, di-iso-butyl phthalate, phthalic acid, diisobutyl ester, phthalic acid diisobutyl ester, di i-butyl phthalate
PubChem CID 6782
ChEBI CHEBI:79053
IUPAC Name 1,2-bis(2-methylpropyl) benzene-1,2-dicarboxylate
SMILES CC(C)COC(=O)C1=CC=CC=C1C(=O)OCC(C)C

Specifications

Color Yellow
Boiling Point 320°C
Quantity 25 g
UN Number 3082
Formula Weight 278.35
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diisobutyl Phthalate
EINECSNumber (3)-1303
RTECSNumber TI1225000
TSCA Yes