missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Dihydralazine sulfate

$419.65 - $797.86

Chemical Identifiers

CAS 7327-87-9
Molecular Formula C8H12N6O4S
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC466231000
View Documents
Thermo Scientific Chemicals
466231000
100 mg
Each for $419.65
Only null left
 
AC466232500
View Documents
Thermo Scientific Chemicals
466232500
250 mg
Each for $797.86
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7327-87-9
C8H12N6O4S

Specifications

7327-87-9
C8H12N6O4S
BWHAMWGGORIDBK-UHFFFAOYSA-N
1,4-dihydrazinylphthalazine; sulfuric acid
Dihydralazine sulfate
100 mg
288.28
OS(O)(=O)=O.NNC1=C2C=CC=CC2=C(NN)N=N1
288.28
Videos
Safety and Handling

Safety and Handling

GHS H Statement Harmful if swallowed.

GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

GHS Signal Word: Warning

EINECSNumber : 230-808-0

SDS
Documents

Documents

RUO – Research Use Only