missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Dihydralazine sulfate

Catalog No. AC466231000
Change view
Click to view available options
Quantity:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466231000 100 mg
AC466232500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466231000 Supplier Thermo Scientific Chemicals Supplier No. 466231000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7327-87-9
Molecular Formula C8H12N6O4S

Specifications

Quantity 100 mg
Formula Weight 288.28
InChI Key BWHAMWGGORIDBK-UHFFFAOYSA-N
SMILES OS(O)(=O)=O.NNC1=C2C=CC=CC2=C(NN)N=N1
IUPAC Name 1,4-dihydrazinylphthalazine; sulfuric acid
Molecular Weight (g/mol) 288.28
Chemical Name or Material Dihydralazine sulfate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

EINECSNumber 230-808-0

RUO – Research Use Only