missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dihexylammonium Acetate (ca. 0.5mol/L in Water) [Ion-Pair Reagent for LC-MS], TCI America™
Specifications
Specifications
| Physical Form | Liquid |
| Color | Colorless |
| pH | 7.4 to 7.8 |
| Quantity | 10 mL |
| Formula Weight | 245.41 |
| Chemical Name or Material | Dihexylammonium Acetate (ca. 0.5mol/L in Water) [Ion-Pair Reagent for LC-MS] |
Safety and Handling
| TSCA | No |