accessibility menu, dialog, popup

UserName

H3CH2NCH3CH3OO

CAS RN 366793-17-1

IUPAC Name: dihexylazanium acetate
Synonyms: dihexylazanium acetate
Molecular Weight (g/mol): 245.41
Molecular Formula: C14H31NO2
InChi Key: VAMSKEFYAOCCKM-UHFFFAOYSA-N
SMILES: CC([O-])=O.CCCCCC[NH2+]CCCCCC
Molecular Weight (g/mol) 
  • (1)
  • (1)
Grade 
  • (1)
Product Line 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results