missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dihexyl Phthalate 98.0+%, TCI America™
$78.29 - $199.46
Chemical Identifiers
| CAS | 84-75-3 |
|---|---|
| Molecular Formula | C20H30O4 |
| Molecular Weight (g/mol) | 334.46 |
| MDL Number | MFCD00043720 |
| InChI Key | KCXZNSGUUQJJTR-UHFFFAOYSA-N |
| Synonym | dihexyl phthalate, di-n-hexyl phthalate, di-n-hexylphthalate, phthalic acid, dihexyl ester, 1,2-benzenedicarboxylic acid, dihexyl ester, phthalic acid di-n-hexyl ester, phthalic acid dihexyl ester, n-dihexyl phthalate, bis n-hexyl phthalate, dihexyl 1,2-benzenedicarboxylate |
| PubChem CID | 6786 |
| ChEBI | CHEBI:34678 |
| IUPAC Name | 1,2-dihexyl benzene-1,2-dicarboxylate |
| SMILES | CCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCC |
Chemical Identifiers
| 84-75-3 | |
| 334.46 | |
| KCXZNSGUUQJJTR-UHFFFAOYSA-N | |
| 6786 | |
| 1,2-dihexyl benzene-1,2-dicarboxylate |
| C20H30O4 | |
| MFCD00043720 | |
| dihexyl phthalate, di-n-hexyl phthalate, di-n-hexylphthalate, phthalic acid, dihexyl ester, 1,2-benzenedicarboxylic acid, dihexyl ester, phthalic acid di-n-hexyl ester, phthalic acid dihexyl ester, n-dihexyl phthalate, bis n-hexyl phthalate, dihexyl 1,2-benzenedicarboxylate | |
| CHEBI:34678 | |
| CCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCC |
Specifications
| 84-75-3 | |
| 333°C | |
| MFCD00043720 | |
| dihexyl phthalate, di-n-hexyl phthalate, di-n-hexylphthalate, phthalic acid, dihexyl ester, 1,2-benzenedicarboxylic acid, dihexyl ester, phthalic acid di-n-hexyl ester, phthalic acid dihexyl ester, n-dihexyl phthalate, bis n-hexyl phthalate, dihexyl 1,2-benzenedicarboxylate | |
| CCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCC | |
| 334.46 | |
| CHEBI:34678 | |
| ≥98.0% (GC) | |
| Dihexyl Phthalate |
| Yellow | |
| C20H30O4 | |
| 5 g | |
| KCXZNSGUUQJJTR-UHFFFAOYSA-N | |
| 1,2-dihexyl benzene-1,2-dicarboxylate | |
| 6786 | |
| 334.46 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-1307
RTECSNumber : TI1100000
TSCA : Yes