missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Difluoromethyl Phenyl Sulfide 98.0+%, TCI America™
$481.30 - $1927.10
Chemical Identifiers
| CAS | 1535-67-7 |
|---|---|
| Molecular Formula | C7H6F2S |
| Molecular Weight (g/mol) | 160.18 |
| MDL Number | MFCD16301013 |
| InChI Key | KDNBCPHMQJEQLV-UHFFFAOYSA-N |
| Synonym | (Difluoromethylthio)benzene, alpha,alpha-Difluorothioanisole |
| PubChem CID | 73752 |
| IUPAC Name | [(difluoromethyl)sulfanyl]benzene |
| SMILES | FC(F)SC1=CC=CC=C1 |
Chemical Identifiers
| 1535-67-7 | |
| 160.18 | |
| KDNBCPHMQJEQLV-UHFFFAOYSA-N | |
| 73752 | |
| FC(F)SC1=CC=CC=C1 |
| C7H6F2S | |
| MFCD16301013 | |
| (Difluoromethylthio)benzene, alpha,alpha-Difluorothioanisole | |
| [(difluoromethyl)sulfanyl]benzene |
Specifications
| 1535-67-7 | |
| 63°C | |
| MFCD16301013 | |
| 1993 | |
| KDNBCPHMQJEQLV-UHFFFAOYSA-N | |
| [(difluoromethyl)sulfanyl]benzene | |
| 73752 | |
| ≥98.0% (GC) | |
| Difluoromethyl Phenyl Sulfide |
| Yellow | |
| C7H6F2S | |
| 1 g | |
| (Difluoromethylthio)benzene, alpha,alpha-Difluorothioanisole | |
| FC(F)SC1=CC=CC=C1 | |
| 160.18 | |
| 160.18 | |
| Liquid |
Safety and Handling
TSCA : No