accessibility menu, dialog, popup

UserName

CAS RN 1535-67-7

FFS

CAS RN 1535-67-7

IUPAC Name: [(difluoromethyl)sulfanyl]benzene
Synonyms: [(difluoromethyl)sulfanyl]benzene
Molecular Weight (g/mol): 160.18
Molecular Formula: C7H6F2S
InChi Key: KDNBCPHMQJEQLV-UHFFFAOYSA-N
SMILES: FC(F)SC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results