missing translation for 'onlineSavingsMsg'
Learn More

Difluoromethyl Phenyl Sulfide 98.0+%, TCI America™

Catalog No. D49381G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49381G 1 g
D49385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49381G Supplier TCI America Supplier No. D49381G
Only null left

Chemical Identifiers

CAS 1535-67-7
Molecular Formula C7H6F2S
Molecular Weight (g/mol) 160.18
MDL Number MFCD16301013
InChI Key KDNBCPHMQJEQLV-UHFFFAOYSA-N
Synonym (Difluoromethylthio)benzene, alpha,alpha-Difluorothioanisole
PubChem CID 73752
IUPAC Name [(difluoromethyl)sulfanyl]benzene
SMILES FC(F)SC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 63°C
Quantity 1 g
UN Number 1993
Formula Weight 160.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Difluoromethyl Phenyl Sulfide