missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Difluoromethoxy)benzene 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 140°C |
| Quantity | 5 g |
| UN Number | 1993 |
| Formula Weight | 144.12 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | (Difluoromethoxy)benzene |
Safety and Handling
| TSCA | No |