accessibility menu, dialog, popup

UserName

FFO

CAS RN 458-92-4

IUPAC Name: (difluoromethoxy)benzene
Synonyms: difluoromethoxybenzene
Molecular Weight (g/mol): 144.12
Molecular Formula: C7H6F2O
InChi Key: LMVBQQAXGZVBFH-UHFFFAOYSA-N
SMILES: FC(F)OC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results