missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Difco™ Soytone

An enzymatic digest of soy.

$231.82 - $3710.25

Specifications

Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Culture Type Mammalian, Insect, Microbial
Form Powder
Product Type Yeast Supplement
For Use With (Application) Mammalian Cell Culture
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
212488
View Documents
Gibco™
212488
500 g
Each for $231.82
Only null left
 
B12489
View Documents
Gibco™
212489
10 kg
Each for $3,710.25
Only null left
 
Description

Description

  • An alternative carbohydrate profile to yeast supplements for mammalian cell processes, such as CHO
  • High concentrations of vitamins and carbohydrates
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
Specifications

Specifications

Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Powder
Mammalian Cell Culture
Animal origin-free
Mammalian, Insect, Microbial
Yeast Supplement
Difco
Videos
SDS
Documents

Documents