missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Noble Agar
Used as a solidifying agent essentially free of impurities. Difco™ Dehydrated Culture Media: Noble Agar is perfect for electrophoretic and nutritional procedures wherever increased purity is required.
$1858.71 - $1858.71
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
|---|---|
| Concentration | 1.5% |
| Content And Storage | <30°C |
| Additive | Noble Agar |
| Form | Dehydrated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
DF0142-17-0
|
BD
214230 |
500 g |
Each for $1,858.71
|
|
|||||
Description
Used as a solidifying agent essentially free of impurities. Difco™ Dehydrated Culture Media: Noble Agar is perfect for electrophoretic and nutritional procedures wherever increased purity is required.
- Solidifying agent
- Free of impurities
Applications:
- Used for electrophoretic and nutritional procedures
Specifications
| ISO/DIS, 11014 | |
| <30°C | |
| Dehydrated | |
| Bottle |
| 1.5% | |
| Noble Agar | |
| Preparing microbiological culture media, for Electrophoretic and nutritional procedures |