missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Noble Agar
Used as a solidifying agent essentially free of impurities. Difco™ Dehydrated Culture Media: Noble Agar is perfect for electrophoretic and nutritional procedures wherever increased purity is required.
Supplier: BD 214230
Description
Used as a solidifying agent essentially free of impurities. Difco™ Dehydrated Culture Media: Noble Agar is perfect for electrophoretic and nutritional procedures wherever increased purity is required.
- Solidifying agent
- Free of impurities
Applications:
- Used for electrophoretic and nutritional procedures
Specifications
| ISO/DIS, 11014 | |
| <30°C | |
| Dehydrated | |
| Bottle |
| 1.5% | |
| Noble Agar | |
| Preparing microbiological culture media, for Electrophoretic and nutritional procedures | |
| 500 g |