missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylstilbestrol 98.0+%, TCI America™
$85.75 - $210.63
Chemical Identifiers
| CAS | 6898-97-1 |
|---|---|
| Molecular Formula | C18H20O2 |
| Molecular Weight (g/mol) | 268.36 |
| MDL Number | MFCD00002373 |
| InChI Key | RGLYKWWBQGJZGM-ISLYRVAYSA-N |
| Synonym | Stilbestrol |
| PubChem CID | 688057 |
| IUPAC Name | 4-[(3E)-4-(4-hydroxyphenyl)hex-3-en-3-yl]phenol |
| SMILES | CC\C(=C(\CC)C1=CC=C(O)C=C1)C1=CC=C(O)C=C1 |
Chemical Identifiers
| 6898-97-1 | |
| 268.36 | |
| RGLYKWWBQGJZGM-ISLYRVAYSA-N | |
| 688057 | |
| CC\C(=C(\CC)C1=CC=C(O)C=C1)C1=CC=C(O)C=C1 |
| C18H20O2 | |
| MFCD00002373 | |
| Stilbestrol | |
| 4-[(3E)-4-(4-hydroxyphenyl)hex-3-en-3-yl]phenol |
Specifications
| 6898-97-1 | |
| White | |
| MFCD00002373 | |
| 3077 | |
| RGLYKWWBQGJZGM-ISLYRVAYSA-N | |
| 4-[(3E)-4-(4-hydroxyphenyl)hex-3-en-3-yl]phenol | |
| 688057 | |
| ≥98.0% (GC) | |
| Diethylstilbestrol |
| 169°C | |
| C18H20O2 | |
| 1 g | |
| Stilbestrol | |
| CC\C(=C(\CC)C1=CC=C(O)C=C1)C1=CC=C(O)C=C1 | |
| 268.36 | |
| 268.36 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : WJ5600000
TSCA : No