accessibility menu, dialog, popup

UserName

CAS RN 6898-97-1

CH3OHCH3OH(E)

CAS RN 6898-97-1

IUPAC Name: 4-[(3E)-4-(4-hydroxyphenyl)hex-3-en-3-yl]phenol
Synonyms: diethylstilbestrol percutatrine oestrogenique iscovesco distilbene desma trans-diethylstilbestrol dietilestilbestrol DEB DES sibol diethylstilboesterol
Molecular Weight (g/mol): 268.36
Molecular Formula: C18H20O2
InChi Key: RGLYKWWBQGJZGM-ISLYRVAYSA-N
SMILES: CC\C(=C(\CC)C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results