missing translation for 'onlineSavingsMsg'
Learn More

Diethylstilbestrol 98.0+%, TCI America™

Catalog No. D05265G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D05265G 5 g
D05261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D05265G Supplier TCI America Supplier No. D05265G
Only null left

Chemical Identifiers

CAS 6898-97-1
Molecular Formula C18H20O2
Molecular Weight (g/mol) 268.36
MDL Number MFCD00002373
InChI Key RGLYKWWBQGJZGM-ISLYRVAYSA-N
Synonym Stilbestrol
PubChem CID 688057
IUPAC Name 4-[(3E)-4-(4-hydroxyphenyl)hex-3-en-3-yl]phenol
SMILES CC\C(=C(\CC)C1=CC=C(O)C=C1)C1=CC=C(O)C=C1

Specifications

Melting Point 169°C
Color White
Quantity 5 g
UN Number 3077
Formula Weight 268.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diethylstilbestrol
RTECSNumber WJ5600000
TSCA No