missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylstilbestrol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 169°C |
| Color | White |
| Quantity | 1 g |
| UN Number | 3077 |
| Formula Weight | 268.36 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Diethylstilbestrol |
Safety and Handling
| RTECSNumber | WJ5600000 |
| TSCA | No |