missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylene Glycol 99.5+%, TCI America™
Supplier: TCI America D0495500G
Specifications
| Diethylene Glycol | |
| -7°C | |
| 245°C | |
| MFCD00002882 | |
| diethylene glycol, 2,2'-oxydiethanol, diglycol, diethylenglykol, 2-hydroxyethyl ether, bis 2-hydroxyethyl ether, ethanol, 2,2'-oxybis, 2,2'-oxybisethanol, 2-2-hydroxyethoxy ethanol, digol | |
| OCCOCCO | |
| 106.12 | |
| CHEBI:46807 | |
| ≥99.5% (GC) |
| 111-46-6 | |
| Yellow | |
| C4H10O3 | |
| 500 g | |
| MTHSVFCYNBDYFN-UHFFFAOYSA-N | |
| 2-(2-hydroxyethoxy)ethan-1-ol | |
| 8117 | |
| 106.12 | |
| Liquid |
Chemical Identifiers
| 111-46-6 | |
| 106.12 | |
| MTHSVFCYNBDYFN-UHFFFAOYSA-N | |
| 8117 | |
| 2-(2-hydroxyethoxy)ethan-1-ol |
| C4H10O3 | |
| MFCD00002882 | |
| diethylene glycol, 2,2'-oxydiethanol, diglycol, diethylenglykol, 2-hydroxyethyl ether, bis 2-hydroxyethyl ether, ethanol, 2,2'-oxybis, 2,2'-oxybisethanol, 2-2-hydroxyethoxy ethanol, digol | |
| CHEBI:46807 | |
| OCCOCCO |
Safety and Handling
EINECSNumber : (2)-0415
RTECSNumber : ID5950000
TSCA : Yes