missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol 99.5+%, TCI America™

Catalog No. p-7126248
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0495100G 100 g
D0495500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0495100G Supplier TCI America Supplier No. D0495100G
Only null left

Chemical Identifiers

CAS 111-46-6
Molecular Formula C4H10O3
Molecular Weight (g/mol) 106.12
MDL Number MFCD00002882
InChI Key MTHSVFCYNBDYFN-UHFFFAOYSA-N
Synonym diethylene glycol, 2,2'-oxydiethanol, diglycol, diethylenglykol, 2-hydroxyethyl ether, bis 2-hydroxyethyl ether, ethanol, 2,2'-oxybis, 2,2'-oxybisethanol, 2-2-hydroxyethoxy ethanol, digol
PubChem CID 8117
ChEBI CHEBI:46807
IUPAC Name 2-(2-hydroxyethoxy)ethan-1-ol
SMILES OCCOCCO

Specifications

Melting Point -7°C
Color Yellow
Boiling Point 245°C
Quantity 100 g
Formula Weight 106.12
Percent Purity ≥99.5% (GC)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol
EINECSNumber (2)-0415
RTECSNumber ID5950000
TSCA Yes