missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylammonium Diethyldithiocarbamate 97.0+%, TCI America™
Supplier: TCI America D0488500G
Specifications
| Diethylammonium Diethyldithiocarbamate | |
| White | |
| Crystalline Powder | |
| C5H11NS2 | |
| Diethyldithiocarbamic Acid Diethylammonium Salt | |
| ≥97.0% (T) |
| 83°C | |
| 1518-58-7 | |
| 500 g | |
| MFCD00013146 | |
| 222.41 |
Chemical Identifiers
| 1518-58-7 | |
| C5H11NS2 | |
| MFCD00013146 | |
| Diethyldithiocarbamic Acid Diethylammonium Salt |
Safety and Handling
EINECSNumber : (2)-2894
RTECSNumber : EZ5425000
TSCA : Yes