missing translation for 'onlineSavingsMsg'
Learn More

Diethylammonium Diethyldithiocarbamate 97.0+%, TCI America™

Catalog No. p-7126243
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D048825G 25 g
D0488500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D048825G Supplier TCI America Supplier No. D048825G
Only null left

Chemical Identifiers

CAS 1518-58-7
Molecular Formula C5H11NS2
MDL Number MFCD00013146
Synonym Diethyldithiocarbamic Acid Diethylammonium Salt

Specifications

Melting Point 83°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
Formula Weight 222.41
Percent Purity ≥97.0% (T)
Chemical Name or Material Diethylammonium Diethyldithiocarbamate
EINECSNumber (2)-2894
RTECSNumber EZ5425000
TSCA Yes