accessibility menu, dialog, popup

UserName

CH3HNH3CH3CNCH3SHS

CAS RN 1518-58-7

IUPAC Name: diethyl[sulfanyl(carbonothioyl)]amine; diethylamine
Synonyms: diethylamine; diethylcarbamodithioic acid
Molecular Weight (g/mol): 222.41
Molecular Formula: C9H22N2S2
InChi Key: NBGTWXBPCIHUQD-UHFFFAOYSA-N
SMILES: CCNCC.CCN(CC)C(S)=S
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)