missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Diethylamino)sulfur Trifluoride 90.0+%, TCI America™
Supplier: TCI America D18685G
Specifications
| (Diethylamino)sulfur Trifluoride [Fluorinating Reagent] | |
| 32°C | |
| MFCD00000363 | |
| 2920 | |
| CSJLBAMHHLJAAS-UHFFFAOYSA-N | |
| N-ethyl-N-(trifluoro-$l^{4}-sulfanyl)ethanamine | |
| 123472 | |
| ≥90.0% (T) |
| 38078-09-0 | |
| C4H10F3NS | |
| 5 g | |
| diethylaminosulfur trifluoride, dast, diethylaminosulphur trifluoride, diethylamino sulfur trifluoride, diethylaminosulfurtrifluoride, diethylaminosulphurtrifluoride, diethylamino sulphur trifluoride, unii-78622bv6ij, sulfur, n-ethylethanaminato trifluoro-, t-4 | |
| CCN(CC)S(F)(F)F | |
| 161.186 | |
| 161.19 | |
| Liquid |
Chemical Identifiers
| 38078-09-0 | |
| 161.186 | |
| CSJLBAMHHLJAAS-UHFFFAOYSA-N | |
| 123472 | |
| CCN(CC)S(F)(F)F |
| C4H10F3NS | |
| MFCD00000363 | |
| diethylaminosulfur trifluoride, dast, diethylaminosulphur trifluoride, diethylamino sulfur trifluoride, diethylaminosulfurtrifluoride, diethylaminosulphurtrifluoride, diethylamino sulphur trifluoride, unii-78622bv6ij, sulfur, n-ethylethanaminato trifluoro-, t-4 | |
| N-ethyl-N-(trifluoro-$l^{4}-sulfanyl)ethanamine |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator