accessibility menu, dialog, popup

UserName

CAS RN 38078-09-0

IUPAC Name: diethyl(trifluoro-λ⁴-sulfanyl)amine
Synonyms: diethylaminosulfur trifluoride
Molecular Weight (g/mol): 161.19
Molecular Formula: C4H10F3NS
InChi Key: CSJLBAMHHLJAAS-UHFFFAOYSA-N
SMILES: CCN(CC)S(F)(F)F
Boiling Point 
  • (3)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (3)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)