missing translation for 'onlineSavingsMsg'
Learn More

(Diethylamino)sulfur Trifluoride 90.0+%, TCI America™

Catalog No. p-7126979
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D18685G 5 g
D186825G 25 g
D1868100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D18685G Supplier TCI America Supplier No. D18685G
Only null left

Chemical Identifiers

CAS 38078-09-0
Molecular Formula C4H10F3NS
Molecular Weight (g/mol) 161.186
MDL Number MFCD00000363
InChI Key CSJLBAMHHLJAAS-UHFFFAOYSA-N
Synonym diethylaminosulfur trifluoride, dast, diethylaminosulphur trifluoride, diethylamino sulfur trifluoride, diethylaminosulfurtrifluoride, diethylaminosulphurtrifluoride, diethylamino sulphur trifluoride, unii-78622bv6ij, sulfur, n-ethylethanaminato trifluoro-, t-4
PubChem CID 123472
IUPAC Name N-ethyl-N-(trifluoro-$l^{4}-sulfanyl)ethanamine
SMILES CCN(CC)S(F)(F)F

Specifications

Boiling Point 32°C
Quantity 5 g
UN Number 2920
Formula Weight 161.19
Percent Purity ≥90.0% (T)
Physical Form Liquid
Chemical Name or Material (Diethylamino)sulfur Trifluoride [Fluorinating Reagent]
TSCA No
Recommended Storage Refrigerator