missing translation for 'onlineSavingsMsg'
Learn More

(Diethylamino)sulfur Trifluoride 90.0+%, TCI America™

Supplier:  TCI America D186825G

 View more versions of this product

Catalog No. D186825G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

(Diethylamino)sulfur Trifluoride [Fluorinating Reagent]
32°C
MFCD00000363
2920
CSJLBAMHHLJAAS-UHFFFAOYSA-N
N-ethyl-N-(trifluoro-$l^{4}-sulfanyl)ethanamine
123472
≥90.0% (T)
38078-09-0
C4H10F3NS
25 g
diethylaminosulfur trifluoride, dast, diethylaminosulphur trifluoride, diethylamino sulfur trifluoride, diethylaminosulfurtrifluoride, diethylaminosulphurtrifluoride, diethylamino sulphur trifluoride, unii-78622bv6ij, sulfur, n-ethylethanaminato trifluoro-, t-4
CCN(CC)S(F)(F)F
161.186
161.19
Liquid

Chemical Identifiers

38078-09-0
161.186
CSJLBAMHHLJAAS-UHFFFAOYSA-N
123472
CCN(CC)S(F)(F)F
C4H10F3NS
MFCD00000363
diethylaminosulfur trifluoride, dast, diethylaminosulphur trifluoride, diethylamino sulfur trifluoride, diethylaminosulfurtrifluoride, diethylaminosulphurtrifluoride, diethylamino sulphur trifluoride, unii-78622bv6ij, sulfur, n-ethylethanaminato trifluoro-, t-4
N-ethyl-N-(trifluoro-$l^{4}-sulfanyl)ethanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions