missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Terephthalate 99.0+%, TCI America™

Catalog No. T001425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T001425G 25 g
T0014500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T001425G Supplier TCI America Supplier No. T001425G

Chemical Identifiers

CAS 636-09-9
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00039891
InChI Key ONIHPYYWNBVMID-UHFFFAOYSA-N
Synonym diethyl terephthalate, p-diethyl phthalate, diethylterephthalate, terephthalic acid, diethyl ester, 1,4-benzenedicarboxylic acid, diethyl ester, diethyl p-phthalate, terephthalic acid diethyl ester, unii-n97x85l3cd, 1,4-diethyl benzene-1,4-dicarboxylate, 1,4-benzenedicarboxylic acid, 1,4-diethyl ester
PubChem CID 12483
IUPAC Name diethyl benzene-1,4-dicarboxylate
SMILES CCOC(=O)C1=CC=C(C=C1)C(=O)OCC

Specifications

Melting Point 45°C
Color White
Boiling Point 142°C
Quantity 25 g
Formula Weight 222.24
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl Terephthalate
EINECSNumber (3)-3618
RTECSNumber WZ1223400
TSCA Yes