accessibility menu, dialog, popup

UserName

H3COOOOCH3

CAS RN 636-09-9

IUPAC Name: 1,4-diethyl benzene-1,4-dicarboxylate
Synonyms: 1,4-diethyl benzene-1,4-dicarboxylate
Molecular Weight (g/mol): 222.24
Molecular Formula: C12H14O4
InChi Key: ONIHPYYWNBVMID-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(C=C1)C(=O)OCC
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)