missing translation for 'onlineSavingsMsg'
Learn More

Diethyl terephthalate, 98%

Catalog No. AC407450250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC407450250 25 g Glass bottle
AC407451000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC407450250 Supplier Thermo Scientific Chemicals Supplier No. 407450250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 636-09-9 | C12H14O4 | 222.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 636-09-9
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00039891
InChI Key ONIHPYYWNBVMID-UHFFFAOYSA-N
Synonym diethyl terephthalate, p-diethyl phthalate, diethylterephthalate, terephthalic acid, diethyl ester, 1,4-benzenedicarboxylic acid, diethyl ester, diethyl p-phthalate, terephthalic acid diethyl ester, unii-n97x85l3cd, 1,4-diethyl benzene-1,4-dicarboxylate, 1,4-benzenedicarboxylic acid, 1,4-diethyl ester
PubChem CID 12483
IUPAC Name diethyl benzene-1,4-dicarboxylate
SMILES CCOC(=O)C1=CC=C(C=C1)C(=O)OCC

Specifications

Melting Point 42.0°C to 45.0°C
Color White
Density 1.1100g/mL
Boiling Point 302.0°C
Flash Point 117°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Specific Gravity 1.11
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol,ether,ethanol
Formula Weight 222.24
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Diethyl terephthalate, 98%
Show More Show Less
EINECSNumber 211-249-1
RTECSNumber WZ1223400
TSCA TSCA

RUO – Research Use Only