missing translation for 'onlineSavingsMsg'
Learn More

Diethyl terephthalate, 95%

Catalog No. p-7025743
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2140109 10 g
AAB2140118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2140109 Supplier Thermo Scientific Chemicals Supplier No. B2140109
Only null left

CAS: 636-09-9 | C12H14O4 | 222.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 636-09-9
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00039891
InChI Key ONIHPYYWNBVMID-UHFFFAOYSA-N
Synonym diethyl terephthalate, p-diethyl phthalate, diethylterephthalate, terephthalic acid, diethyl ester, 1,4-benzenedicarboxylic acid, diethyl ester, diethyl p-phthalate, terephthalic acid diethyl ester, unii-n97x85l3cd, 1,4-diethyl benzene-1,4-dicarboxylate, 1,4-benzenedicarboxylic acid, 1,4-diethyl ester
PubChem CID 12483
IUPAC Name diethyl benzene-1,4-dicarboxylate
SMILES CCOC(=O)C1=CC=C(C=C1)C(=O)OCC

Specifications

Melting Point 44°C to 46°C
Density 1.1
Quantity 10 g
Formula Weight 222.24
Percent Purity 95%
Chemical Name or Material Diethyl terephthalate
EINECSNumber 211-249-1
RTECSNumber WZ1223400
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only