missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Mesoxalate 94.0+%, TCI America™
Supplier: TCI America M086625G
Specifications
| Diethyl Mesoxalate | |
| 210°C | |
| MFCD00009121 | |
| diethyl ketomalonate, diethyl mesoxalate, diethyl 2-oxomalonate, diethyl oxomalonate, ethyl mesoxalate, ethyl ketomalonate, propanedioic acid, oxo-, diethyl ester, diethylketomalonate, mesoxalic acid diethyl ester, mesoxalic acid, diethyl ester | |
| CCOC(=O)C(=O)C(=O)OCC | |
| 174.152 | |
| 174.15 | |
| Liquid |
| 609-09-6 | |
| C7H10O5 | |
| 25 g | |
| DBKKFIIYQGGHJO-UHFFFAOYSA-N | |
| diethyl 2-oxopropanedioate | |
| 69105 | |
| ≥94.0% (GC) |
Chemical Identifiers
| 609-09-6 | |
| 174.152 | |
| DBKKFIIYQGGHJO-UHFFFAOYSA-N | |
| 69105 | |
| CCOC(=O)C(=O)C(=O)OCC |
| C7H10O5 | |
| MFCD00009121 | |
| diethyl ketomalonate, diethyl mesoxalate, diethyl 2-oxomalonate, diethyl oxomalonate, ethyl mesoxalate, ethyl ketomalonate, propanedioic acid, oxo-, diethyl ester, diethylketomalonate, mesoxalic acid diethyl ester, mesoxalic acid, diethyl ester | |
| diethyl 2-oxopropanedioate |
Safety and Handling
TSCA : Yes