accessibility menu, dialog, popup

UserName

H3COOOOOCH3

CAS RN 609-09-6

IUPAC Name: 1,3-diethyl 2-oxopropanedioate
Synonyms: 1,3-diethyl 2-oxopropanedioate
Molecular Weight (g/mol): 174.15
Molecular Formula: C7H10O5
InChi Key: DBKKFIIYQGGHJO-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=O)C(=O)OCC
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)