missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Mesoxalate 94.0+%, TCI America™

Catalog No. p-7132413
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08665G 5 g
M086625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08665G Supplier TCI America Supplier No. M08665G
Only null left

Chemical Identifiers

CAS 609-09-6
Molecular Formula C7H10O5
Molecular Weight (g/mol) 174.152
MDL Number MFCD00009121
InChI Key DBKKFIIYQGGHJO-UHFFFAOYSA-N
Synonym diethyl ketomalonate, diethyl mesoxalate, diethyl 2-oxomalonate, diethyl oxomalonate, ethyl mesoxalate, ethyl ketomalonate, propanedioic acid, oxo-, diethyl ester, diethylketomalonate, mesoxalic acid diethyl ester, mesoxalic acid, diethyl ester
PubChem CID 69105
IUPAC Name diethyl 2-oxopropanedioate
SMILES CCOC(=O)C(=O)C(=O)OCC

Specifications

Boiling Point 210°C
Quantity 5 g
Formula Weight 174.15
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Mesoxalate