missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl iminodicarboxylate, 98%
CAS: 19617-44-8 | C6H11NO4 | 161.16 g/mol
Supplier: Thermo Scientific Chemicals 460770050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 51.0°C to 53.0°C | |
| 19617-44-8 | |
| Authentic | |
| C6H11NO4 | |
| PQVSTLUFSYVLTO-UHFFFAOYSA-N | |
| ethyl N-ethoxycarbonylcarbamate | |
| 161.16 | |
| 161.16 |
| Diethyl iminodicarboxylate | |
| 132.0°C to 134.0°C (15.0mmHg) | |
| 97.5% min. (GC) | |
| azamalonic ester, diethyl imidodicarbonate, diethyl azamalonate, imidodicarbonic acid, diethyl ester, diethyl iminodicarboxylate, diethylazamalonate, imidodicarbonic acid,1,3-diethyl ester, diethylimidodicarbonate, diethyl imidodicarbonate #, ethyl ethoxycarbonylamino formate | |
| CCOC(=O)NC(=O)OCC | |
| 5 g | |
| 140564 | |
| 98% |
Chemical Identifiers
| 19617-44-8 | |
| 161.16 | |
| azamalonic ester, diethyl imidodicarbonate, diethyl azamalonate, imidodicarbonic acid, diethyl ester, diethyl iminodicarboxylate, diethylazamalonate, imidodicarbonic acid,1,3-diethyl ester, diethylimidodicarbonate, diethyl imidodicarbonate #, ethyl ethoxycarbonylamino formate | |
| ethyl N-ethoxycarbonylcarbamate |
| C6H11NO4 | |
| PQVSTLUFSYVLTO-UHFFFAOYSA-N | |
| 140564 | |
| CCOC(=O)NC(=O)OCC |
RUO – Research Use Only