missing translation for 'onlineSavingsMsg'
Learn More

Diethyl iminodicarboxylate, 98%

Catalog No. p-7077855
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460770010 1 g
AC460770050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC460770010 Supplier Thermo Scientific Chemicals Supplier No. 460770010

CAS: 19617-44-8 | C6H11NO4 | 161.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19617-44-8
Molecular Formula C6H11NO4
Molecular Weight (g/mol) 161.16
InChI Key PQVSTLUFSYVLTO-UHFFFAOYSA-N
Synonym azamalonic ester, diethyl imidodicarbonate, diethyl azamalonate, imidodicarbonic acid, diethyl ester, diethyl iminodicarboxylate, diethylazamalonate, imidodicarbonic acid,1,3-diethyl ester, diethylimidodicarbonate, diethyl imidodicarbonate #, ethyl ethoxycarbonylamino formate
PubChem CID 140564
IUPAC Name ethyl N-ethoxycarbonylcarbamate
SMILES CCOC(=O)NC(=O)OCC

Specifications

Melting Point 51.0°C to 53.0°C
Boiling Point 132.0°C to 134.0°C (15.0mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 161.16
Percent Purity 98%
Chemical Name or Material Diethyl iminodicarboxylate

RUO – Research Use Only