missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Ethyl(phenyl)malonate 98.0+%, TCI America™

Catalog No. p-7127700
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33755G 5 g
D337525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33755G Supplier TCI America Supplier No. D33755G

Chemical Identifiers

CAS 76-67-5
Molecular Formula C15H20O4
Molecular Weight (g/mol) 264.321
MDL Number MFCD00040759
InChI Key PKRVDBARWFJWEB-UHFFFAOYSA-N
Synonym diethyl 2-ethyl-2-phenylmalonate, diethyl ethylphenylmalonate, diethyl ethyl phenyl malonate, propanedioic acid, ethylphenyl-, diethyl ester, ethylphenylmalonic acid diethyl ester, diethyl 2-ethyl-2-phenyl-propanedioate, propanedioic acid, 2-ethyl-2-phenyl-, 1,3-diethyl ester, 1,3-diethyl 2-ethyl-2-phenylpropanedioate, diethyl 2-ethyl-2-phenylpropane-1,3-dioate, malonic acid, ethylphenyl-, diethyl ester
PubChem CID 66450
IUPAC Name diethyl 2-ethyl-2-phenylpropanedioate
SMILES CCC(C1=CC=CC=C1)(C(=O)OCC)C(=O)OCC

Specifications

Melting Point -7°C
Color Yellow
Boiling Point 171°C
Quantity 5 g
Formula Weight 264.32
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Ethyl(phenyl)malonate