accessibility menu, dialog, popup

UserName

CH3OOH3COOH3C

CAS RN 76-67-5

IUPAC Name: 1,3-diethyl 2-ethyl-2-phenylpropanedioate
Synonyms: 1,3-diethyl 2-ethyl-2-phenylpropanedioate
Molecular Weight (g/mol): 264.32
Molecular Formula: C15H20O4
InChi Key: PKRVDBARWFJWEB-UHFFFAOYSA-N
SMILES: CCOC(=O)C(CC)(C(=O)OCC)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results