missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Disulfide 99.0+%, TCI America™
Supplier: TCI America D048625G
Specifications
Diethyl Disulfide | |
Yellow | |
C4H10S2 | |
25 g | |
diethyl disulfide, ethyl disulfide, disulfide, diethyl, diethyl disulphide, ethyldithioethane, 3,4-dithiahexane, ethyl disulphide, ethyldisulfanyl ethane, diethyldisulfid, diethyldisulfide | |
CCSSCC | |
122.244 | |
122.24 | |
Liquid |
110-81-6 | |
154°C | |
MFCD00009266 | |
1993 | |
CETBSQOFQKLHHZ-UHFFFAOYSA-N | |
(ethyldisulfanyl)ethane | |
8077 | |
≥99.0% (GC) |
Chemical Identifiers
110-81-6 | |
122.244 | |
CETBSQOFQKLHHZ-UHFFFAOYSA-N | |
8077 | |
CCSSCC |
C4H10S2 | |
MFCD00009266 | |
diethyl disulfide, ethyl disulfide, disulfide, diethyl, diethyl disulphide, ethyldithioethane, 3,4-dithiahexane, ethyl disulphide, ethyldisulfanyl ethane, diethyldisulfid, diethyldisulfideShow More | |
(ethyldisulfanyl)ethane |
Safety and Handling
EINECSNumber : (2)-0477&(2)-2421
RTECSNumber : JO1925000
TSCA : Yes