accessibility menu, dialog, popup

UserName

CAS RN 110-81-6

IUPAC Name: (ethyldisulfanyl)ethane
Synonyms: diethyl disulfide
Molecular Weight (g/mol): 122.24
Molecular Formula: C4H10S2
InChi Key: CETBSQOFQKLHHZ-UHFFFAOYSA-N
SMILES: CCSSCC
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)