missing translation for 'onlineSavingsMsg'
Learn More

Diethyl disulfide, 99%

Catalog No. AAL1006514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1006514 25 g
AAL1006522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1006514 Supplier Thermo Scientific Chemicals Supplier No. L1006514

CAS: 110-81-6 | C4H10S2 | 122.244 g/mol

Diethyl sulfide is used as a solvent for anhydrous minerals and in plating baths for gold and silver

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl sulfide is used as a solvent for anhydrous minerals and in plating baths for gold and silver

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 110-81-6
Molecular Formula C4H10S2
Molecular Weight (g/mol) 122.244
MDL Number MFCD00009266
InChI Key CETBSQOFQKLHHZ-UHFFFAOYSA-N
Synonym diethyl disulfide, ethyl disulfide, disulfide, diethyl, diethyl disulphide, ethyldithioethane, 3,4-dithiahexane, ethyl disulphide, ethyldisulfanyl ethane, diethyldisulfid, diethyldisulfide
PubChem CID 8077
IUPAC Name (ethyldisulfanyl)ethane
SMILES CCSSCC

Specifications

Melting Point -102°C
Density 0.993
Boiling Point 152°C to 154°C
Flash Point 40°C (104°F)
Odor Stench
Refractive Index 1.506
Linear Formula CH3CH2S-SCH2CH3
Quantity 25 g
UN Number UN1993
Beilstein 1098273
Solubility Information Soluble in water.
Formula Weight 122.25
Percent Purity 99%
Chemical Name or Material Diethyl disulfide
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H319-H335
Flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 203-805-7
RTECSNumber JO1925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only